Assay Details
| Gene Symbol: emb2411 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198014.1 | NP_001184943.1 | 11 - 12 | 1586 | 139 | |
| NM_001331794.1 | NP_001318959.1 | 13 - 14 | 1748 | 139 | ||
| NM_001331795.1 | NP_001322072.1 | 13 - 14 | 1792 | 139 | ||
| NM_001331796.1 | NP_001322073.1 | 13 - 14 | 1786 | 139 | ||
| NM_001331797.1 | NP_001322071.1 | 11 - 12 | 1586 | 139 | ||
More Information
Set Membership: |
|

emb2411
Genomic Map