Assay Details
| Gene Symbol: emb2411 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198014.1 | NP_001184943.1 | 21 - 22 | 2673 | 127 | |
| NM_001331794.1 | NP_001318959.1 | 22 - 23 | 2778 | 127 | ||
| NM_001331795.1 | NP_001322072.1 | 23 - 24 | 2879 | 127 | ||
| NM_001331796.1 | NP_001322073.1 | 23 - 24 | 2873 | 127 | ||
| NM_001331797.1 | NP_001322071.1 | 21 - 22 | 2673 | 127 | ||
More Information
Set Membership: |
|

emb2411
Genomic Map