Assay Details
| Gene Symbol: emb2411 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198014.1 | NP_001184943.1 | 31 - 32 | 3669 | 124 | |
| NM_001331794.1 | NP_001318959.1 | 32 - 33 | 3774 | 124 | ||
| NM_001331795.1 | NP_001322072.1 | 33 - 34 | 3875 | 124 | ||
| NM_001331796.1 | NP_001322073.1 | 33 - 34 | 3869 | 124 | ||
More Information
Set Membership: |
|

emb2411
Genomic Map