Assay Details
| Gene Symbol: emb2411 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198014.1 | NP_001184943.1 | 6 - 7 | 1107 | 70 | |
| NM_001331794.1 | NP_001318959.1 | 8 - 9 | 1269 | 70 | ||
| NM_001331795.1 | NP_001322072.1 | 8 - 9 | 1313 | 70 | ||
| NM_001331796.1 | NP_001322073.1 | 8 - 9 | 1307 | 70 | ||
| NM_001331797.1 | NP_001322071.1 | 6 - 7 | 1107 | 70 | ||
More Information
Set Membership: |
|

emb2411
Genomic Map