Assay Details
| Gene Symbol: AT1G08940 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100766.3 | NP_172369.1 | 1 - 2 | 722 | 68 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT1G08940| Gene Symbol: AT1G08940 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100766.3 | NP_172369.1 | 1 - 2 | 722 | 68 | |
Set Membership: |
|