Assay Details
| Gene Symbol: SNF4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100773.4 | NP_563834.1 | 11 - 12 | 1905 | 73 | |
| GenBank mRNA | AF439826.1 | - | 11 - 12 | 1858 | 73 | |
| BT000625.1 | - | 11 - 12 | 1255 | 73 | ||
| DQ132632.1 | - | 11 - 12 | 1858 | 73 | ||
More Information
Set Membership: |
|

SNF4
Genomic Map