Assay Details
| Gene Symbol: iPGAM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100850.3 | NP_563852.1 | 1 - 2 | 270 | 89 | |
| GenBank mRNA | AY086182.1 | - | 1 - 2 | 215 | 89 | |
| AY150432.1 | - | 1 - 2 | 158 | 89 | ||
| BT000773.1 | - | 1 - 2 | 215 | 89 | ||
More Information
Set Membership: |
|

iPGAM1
Genomic Map