Assay Details
| Gene Symbol: iPGAM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100850.3 | NP_563852.1 | 4 - 5 | 1368 | 80 | |
| GenBank mRNA | AY086182.1 | - | 4 - 5 | 1313 | 80 | |
| AY150432.1 | - | 4 - 5 | 1256 | 80 | ||
| BT000773.1 | - | 4 - 5 | 1313 | 80 | ||
More Information
Set Membership: |
|

iPGAM1
Genomic Map