Assay Details
| Gene Symbol: BCAT-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035938.2 | NP_001031015.1 | 9 - 10 | 1019 | 105 | |
| NM_001035939.3 | NP_001031016.2 | 8 - 9 | 1129 | 105 | ||
| NM_100881.3 | NP_172478.1 | 8 - 9 | 1103 | 105 | ||
| GenBank mRNA | AF370135.1 | - | 8 - 9 | 1100 | 105 | |
| AJ271731.1 | - | 8 - 9 | 987 | 105 | ||
| AK317528.1 | - | 9 - 10 | 1020 | 105 | ||
| AK319062.1 | - | 8 - 9 | 1126 | 105 | ||
| AY051038.1 | - | 8 - 9 | 935 | 105 | ||
More Information
Set Membership: |
|

BCAT-2
Genomic Map