Assay Details
| Gene Symbol: RECQ4A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100968.3 | NP_172562.2 | 17 - 18 | 2643 | 78 | |
| GenBank mRNA | AY120761.1 | - | 17 - 18 | 2541 | 78 | |
| BT010133.1 | - | 16 - 17 | 2337 | 78 | ||
More Information
Set Membership: |
|

RECQ4A
Genomic Map