Assay Details
| Gene Symbol: XBAT32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125157.3 | NP_200582.3 | 1 - 2 | 703 | 62 | |
| GenBank mRNA | BT011688.1 | - | 1 - 2 | 575 | 62 | |
| BT012272.1 | - | 1 - 2 | 198 | 62 | ||
More Information
Set Membership: |
|

XBAT32
Genomic Map