Assay Details
| Gene Symbol: XBAT32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125157.3 | NP_200582.3 | 6 - 7 | 1215 | 70 | |
| GenBank mRNA | BT011688.1 | - | 6 - 7 | 1087 | 70 | |
| BT012272.1 | - | 6 - 7 | 710 | 70 | ||
More Information
Set Membership: |
|

XBAT32
Genomic Map