Assay Details
| Gene Symbol: NRPB7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125308.4 | NP_200726.1 | 4 - 5 | 541 | 102 | |
| GenBank mRNA | AK118952.1 | - | 4 - 5 | 480 | 102 | |
| BT005667.1 | - | 4 - 5 | 398 | 102 | ||
More Information
Set Membership: |
|

NRPB7
Genomic Map