Assay Details
| Gene Symbol: TOE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203647.1 | NP_001190576.1 | 6 - 7 | 1376 | 61 | |
| NM_001345390.1 | NP_001331668.1 | 6 - 7 | 1376 | 61 | ||
| NM_001345391.1 | NP_001331666.1 | 6 - 7 | 1376 | 61 | ||
| NM_001345392.1 | NP_001331665.1 | 6 - 7 | 1376 | 61 | ||
| NM_001345393.1 | NP_001331667.1 | 6 - 7 | 1376 | 61 | ||
| NM_125405.4 | NP_200820.3 | 6 - 7 | 1376 | 61 | ||
| GenBank mRNA | BT001916.1 | - | 6 - 7 | 912 | 61 | |
More Information
Set Membership: |
|

TOE2
Genomic Map