Assay Details
| Gene Symbol: TOE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125405.4 | NP_200820.3 | 7 - 8 | 1509 | 123 | |
| GenBank mRNA | BT001916.1 | - | 7 - 8 | 1045 | 123 | |
More Information
Set Membership: |
|

TOE2
Genomic Map