Assay Details
| Gene Symbol: TOE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203647.1 | NP_001190576.1 | 9 - 10 | 2021 | 66 | |
| NM_001345391.1 | NP_001331666.1 | 8 - 9 | 2338 | 66 | ||
| NM_001345392.1 | NP_001331665.1 | 8 - 9 | 2155 | 66 | ||
| NM_001345393.1 | NP_001331667.1 | 9 - 10 | 1934 | 66 | ||
| NM_125405.4 | NP_200820.3 | 9 - 10 | 1955 | 66 | ||
More Information
Set Membership: |
|

TOE2
Genomic Map