Assay Details
| Gene Symbol: AGL8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001085307.1 | NP_001078776.1 | 2 - 3 | 93 | 65 | |
| NM_125484.4 | NP_568929.1 | 2 - 3 | 518 | 65 | ||
| GenBank mRNA | AF386929.1 | - | 2 - 3 | 321 | 65 | |
| AY072463.1 | - | 2 - 3 | 262 | 65 | ||
| U33473.1 | - | 2 - 3 | 362 | 65 | ||
More Information
Set Membership: |
|

AGL8
Genomic Map