Assay Details
| Gene Symbol: AT5G60930 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001345436.1 | NP_001332385.1 | 1 - 2 | 339 | 103 | |
| NM_001345437.1 | NP_001332386.1 | 1 - 2 | 339 | 103 | ||
| NM_125486.3 | NP_200901.2 | 1 - 2 | 304 | 103 | ||
More Information
Set Membership: |
|

AT5G60930
Genomic Map