Assay Details
| Gene Symbol: Tudor2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125572.3 | NP_200986.1 | 7 - 8 | 1206 | 129 | |
| GenBank mRNA | BT002044.1 | - | 7 - 8 | 1152 | 129 | |
More Information
Set Membership: |
|

Tudor2
Genomic Map