Assay Details
| Gene Symbol: UBC20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_103932.4 | NP_564572.1 | 1 - 2 | 472 | 67 | |
| GenBank mRNA | AK227382.1 | - | 1 - 2 | 290 | 67 | |
| AY127574.1 | - | 1 - 2 | 266 | 67 | ||
| BT005855.1 | - | 1 - 2 | 118 | 67 | ||
| DQ027034.1 | - | 1 - 2 | 118 | 67 | ||
More Information
Set Membership: |
|

UBC20
Genomic Map