Assay Details
| Gene Symbol: AT1G50670 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001333447.1 | NP_001321194.1 | 3 - 4 | 199 | 130 | |
| NM_001333448.1 | NP_001321195.1 | 3 - 4 | 158 | 130 | ||
| NM_103949.4 | NP_175482.1 | 3 - 4 | 354 | 130 | ||
| GenBank mRNA | BT002977.1 | - | 3 - 4 | 306 | 130 | |
| BT004408.1 | - | 2 - 3 | 110 | 130 | ||
| JQ013443.1 | - | 2 - 3 | 110 | 130 | ||
More Information
Set Membership: |
|

AT1G50670
Genomic Map