Assay Details
| Gene Symbol: AT1G51130 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_103992.3 | NP_175525.1 | 6 - 7 | 1120 | 118 | |
| GenBank mRNA | BT004096.1 | - | 6 - 7 | 1106 | 118 | |
| BT006087.1 | - | 6 - 7 | 961 | 118 | ||
More Information
Set Membership: |
|

AT1G51130
Genomic Map