Assay Details
| Gene Symbol: GRF6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036791.2 | NP_001031868.1 | 2 - 3 | 689 | 114 | |
| NM_001203346.1 | NP_001190275.1 | 2 - 3 | 689 | 114 | ||
| NM_001203347.1 | NP_001190276.1 | 2 - 3 | 689 | 114 | ||
| NM_121083.4 | NP_568229.1 | 2 - 3 | 879 | 114 | ||
| GenBank mRNA | AY052293.1 | - | 2 - 3 | 659 | 114 | |
| AY061918.1 | - | 2 - 3 | 603 | 114 | ||
| U02565.1 | - | 2 - 3 | 637 | 114 | ||
| U68545.1 | - | 2 - 3 | 657 | 114 | ||
More Information
Set Membership: |
|

GRF6
Genomic Map