Assay Details
| Gene Symbol: GRF6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121083.4 | NP_568229.1 | 3 - 4 | 1005 | 111 | |
| GenBank mRNA | AY052293.1 | - | 3 - 4 | 785 | 111 | |
| U02565.1 | - | 3 - 4 | 763 | 111 | ||
| U68545.1 | - | 3 - 4 | 783 | 111 | ||
More Information
Set Membership: |
|

GRF6
Genomic Map