Assay Details
| Gene Symbol: MSL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001085098.2 | NP_001078567.1 | 9 - 10 | 939 | 85 | |
| NM_001203349.1 | NP_001190278.1 | 9 - 10 | 1047 | 85 | ||
| NM_121087.3 | NP_568230.2 | 9 - 10 | 1085 | 85 | ||
| GenBank mRNA | AK221831.1 | - | 9 - 10 | 999 | 85 | |
| AK230015.1 | - | 9 - 10 | 968 | 85 | ||
| AK317509.1 | - | 9 - 10 | 900 | 85 | ||
More Information
Set Membership: |
|

MSL2
Genomic Map