Assay Details
| Gene Symbol: MSL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001085098.2 | NP_001078567.1 | 11 - 12 | 1218 | 73 | |
| NM_001203349.1 | NP_001190278.1 | 11 - 12 | 1326 | 73 | ||
| NM_121087.3 | NP_568230.2 | 11 - 12 | 1364 | 73 | ||
| GenBank mRNA | AK221831.1 | - | 11 - 12 | 1278 | 73 | |
| AK230015.1 | - | 11 - 12 | 1247 | 73 | ||
| AK317509.1 | - | 11 - 12 | 1179 | 73 | ||
More Information
Set Membership: |
|

MSL2
Genomic Map