Assay Details
| Gene Symbol: ASP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121190.3 | NP_196713.1 | 3 - 4 | 452 | 131 | |
| GenBank mRNA | AY050765.1 | - | 3 - 4 | 362 | 131 | |
| AY079310.1 | - | 3 - 4 | 319 | 131 | ||
| U15034.1 | - | 3 - 4 | 362 | 131 | ||
More Information
Set Membership: |
|

ASP3
Genomic Map