Assay Details
| Gene Symbol: AT5G12900 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121293.3 | NP_196794.1 | 1 - 2 | 386 | 65 | |
| GenBank mRNA | AB177845.1 | - | 1 - 2 | 359 | 65 | |
| AY140032.1 | - | 1 - 2 | 384 | 65 | ||
| BT008908.1 | - | 1 - 2 | 212 | 65 | ||
More Information
Set Membership: |
|

AT5G12900
Genomic Map