Assay Details
| Gene Symbol: emb1345 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036344.1 | NP_001031421.1 | 6 - 7 | 862 | 69 | |
| NM_128165.4 | NP_565615.1 | 6 - 7 | 940 | 69 | ||
| GenBank mRNA | AX765828.1 | - | 6 - 7 | 763 | 69 | |
| AY085277.1 | - | 6 - 7 | 862 | 69 | ||
| AY093154.1 | - | 6 - 7 | 827 | 69 | ||
| BT003394.1 | - | 6 - 7 | 763 | 69 | ||
More Information
Set Membership: |
|

emb1345
Genomic Map