Assay Details
| Gene Symbol: KT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_128222.6 | NP_180233.1 | 1 - 2 | 466 | 92 | |
| GenBank mRNA | AK317729.1 | - | 1 - 2 | 237 | 92 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

KT1| Gene Symbol: KT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_128222.6 | NP_180233.1 | 1 - 2 | 466 | 92 | |
| GenBank mRNA | AK317729.1 | - | 1 - 2 | 237 | 92 | |
Set Membership: |
|