Assay Details
| Gene Symbol: KT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001336078.1 | NP_001324538.1 | 3 - 4 | 947 | 133 | |
| NM_128222.6 | NP_180233.1 | 4 - 5 | 1202 | 133 | ||
| GenBank mRNA | AK317729.1 | - | 4 - 5 | 973 | 133 | |
| BT006442.1 | - | 3 - 4 | 593 | 133 | ||
| X62907.1 | - | 965 | 133 | |||
More Information
Set Membership: |
|

KT1
Genomic Map