Assay Details
| Gene Symbol: PD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_128342.3 | NP_180350.1 | 1 | 1268 | 87 | |
| GenBank mRNA | BT025989.1 | - | 1 | 1031 | 87 | |
| DQ411464.1 | - | 1 | 1035 | 87 | ||
More Information
Set Membership: |
|

PD1
Genomic Map