Assay Details
| Gene Symbol: AT2G28120 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_128372.3 | NP_180379.1 | 1 | 1858 | 112 | |
| GenBank mRNA | AY057618.1 | - | 1 | 1857 | 112 | |
| AY062537.1 | - | 1 | 1750 | 112 | ||
| AY093327.1 | - | 1 | 1611 | 112 | ||
More Information
Set Membership: |
|

AT2G28120
Genomic Map