Assay Details
| Gene Symbol: BGLU19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113032.4 | NP_188774.2 | 1 - 2 | 255 | 76 | |
| GenBank mRNA | AY064046.1 | - | 1 - 2 | 230 | 76 | |
| AY096383.1 | - | 1 - 2 | 157 | 76 | ||
More Information
Set Membership: |
|

BGLU19
Genomic Map