Assay Details
| Gene Symbol: BGLU19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113032.4 | NP_188774.2 | 3 - 4 | 379 | 107 | |
| GenBank mRNA | AY064046.1 | - | 3 - 4 | 354 | 107 | |
| AY096383.1 | - | 3 - 4 | 281 | 107 | ||
More Information
Set Membership: |
|

BGLU19
Genomic Map