Assay Details
| Gene Symbol: BGLU19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113032.4 | NP_188774.2 | 5 - 6 | 539 | 126 | |
| GenBank mRNA | AY064046.1 | - | 5 - 6 | 514 | 126 | |
| AY096383.1 | - | 5 - 6 | 441 | 126 | ||
More Information
Set Membership: |
|

BGLU19
Genomic Map