Assay Details
| Gene Symbol: BGLU19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113032.4 | NP_188774.2 | 9 - 10 | 1213 | 76 | |
| GenBank mRNA | AY064046.1 | - | 9 - 10 | 1188 | 76 | |
| AY096383.1 | - | 9 - 10 | 1115 | 76 | ||
More Information
Set Membership: |
|

BGLU19
Genomic Map