Assay Details
| Gene Symbol: RPS10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113128.3 | NP_188869.1 | 1 - 2 | 436 | 115 | |
| GenBank mRNA | AF370548.1 | - | 1 - 2 | 405 | 115 | |
| BT006286.1 | - | 1 - 2 | 336 | 115 | ||
More Information
Set Membership: |
|

RPS10
Genomic Map