Assay Details
| Gene Symbol: TIF3C2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113186.4 | NP_188926.4 | 2 - 3 | 1649 | 64 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TIF3C2| Gene Symbol: TIF3C2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113186.4 | NP_188926.4 | 2 - 3 | 1649 | 64 | |
Set Membership: |
|