Assay Details
| Gene Symbol: SYP32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203032.1 | NP_001189961.1 | 3 - 4 | 666 | 125 | |
| NM_113342.4 | NP_189078.2 | 2 - 3 | 690 | 125 | ||
| GenBank mRNA | BT004300.1 | - | 2 - 3 | 590 | 125 | |
| BT008604.1 | - | 2 - 3 | 499 | 125 | ||
More Information
Set Membership: |
|

SYP32
Genomic Map