Assay Details
| Gene Symbol: SYP32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203032.1 | NP_001189961.1 | 5 - 6 | 1053 | 86 | |
| NM_113342.4 | NP_189078.2 | 4 - 5 | 1077 | 86 | ||
| GenBank mRNA | BT004300.1 | - | 4 - 5 | 977 | 86 | |
| BT008604.1 | - | 4 - 5 | 886 | 86 | ||
More Information
Set Membership: |
|

SYP32
Genomic Map