Assay Details
| Gene Symbol: UBC37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001338687.1 | NP_001325555.1 | 3 - 4 | 748 | 125 | |
| NM_001338688.1 | NP_001325554.1 | 4 - 5 | 701 | 125 | ||
| NM_001338689.1 | NP_001325556.1 | 4 - 5 | 692 | 125 | ||
| NM_113362.2 | NP_566751.1 | 5 - 6 | 607 | 125 | ||
More Information
Set Membership: |
|

UBC37
Genomic Map