Assay Details
| Gene Symbol: AT3G24670 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113378.2 | NP_189110.1 | 4 - 5 | 854 | 111 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT3G24670| Gene Symbol: AT3G24670 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113378.2 | NP_189110.1 | 4 - 5 | 854 | 111 | |
Set Membership: |
|