Assay Details
| Gene Symbol: RPL15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113496.5 | NP_189221.1 | 3 - 4 | 692 | 96 | |
| GenBank mRNA | AF370235.1 | - | 3 - 4 | 620 | 96 | |
| AF424601.1 | - | 3 - 4 | 620 | 96 | ||
| AY059125.1 | - | 3 - 4 | 595 | 96 | ||
| AY084483.1 | - | 3 - 4 | 655 | 96 | ||
More Information
Set Membership: |
|

RPL15
Genomic Map