Assay Details
| Gene Symbol: ARP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113614.3 | NP_189336.1 | 6 - 7 | 689 | 118 | |
| GenBank mRNA | AF507910.1 | - | 6 - 7 | 491 | 118 | |
More Information
Set Membership: |
|

ARP2
Genomic Map