Assay Details
| Gene Symbol: ADK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037052.2 | NP_001032129.1 | 2 - 3 | 356 | 90 | |
| NM_125735.4 | NP_201145.1 | 2 - 3 | 356 | 90 | ||
| GenBank mRNA | AY039945.1 | - | 2 - 3 | 280 | 90 | |
| AY079340.1 | - | 2 - 3 | 266 | 90 | ||
| AY085188.1 | - | 2 - 3 | 313 | 90 | ||
More Information
Set Membership: |
|

ADK1
Genomic Map