Assay Details
| Gene Symbol: BGAL10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125776.2 | NP_201186.1 | 4 - 5 | 625 | 111 | |
| GenBank mRNA | AY093118.1 | - | 4 - 5 | 625 | 111 | |
| BT010338.1 | - | 4 - 5 | 457 | 111 | ||
More Information
Set Membership: |
|

BGAL10
Genomic Map