Assay Details
| Gene Symbol: CHR24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001345616.1 | NP_001332133.1 | 8 - 9 | 1876 | 75 | |
| NM_125791.3 | NP_201200.2 | 8 - 9 | 1978 | 75 | ||
| GenBank mRNA | AF462829.1 | - | 8 - 9 | 1812 | 75 | |
| AY142010.1 | - | 8 - 9 | 1712 | 75 | ||
More Information
Set Membership: |
|

CHR24
Genomic Map