Assay Details
| Gene Symbol: CHR24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001345616.1 | NP_001332133.1 | 9 - 10 | 1964 | 109 | |
| NM_125791.3 | NP_201200.2 | 9 - 10 | 2066 | 109 | ||
| GenBank mRNA | AF462829.1 | - | 9 - 10 | 1900 | 109 | |
| AY142010.1 | - | 9 - 10 | 1800 | 109 | ||
More Information
Set Membership: |
|

CHR24
Genomic Map