Assay Details
| Gene Symbol: AGL78 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125931.2 | NP_201336.1 | 1 | 893 | 101 | |
| GenBank mRNA | AY141224.1 | - | 1 | 893 | 101 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AGL78| Gene Symbol: AGL78 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125931.2 | NP_201336.1 | 1 | 893 | 101 | |
| GenBank mRNA | AY141224.1 | - | 1 | 893 | 101 | |
Set Membership: |
|